Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dichloromethylvinylsilane

Catalog No. 104914500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-491-4500G 500 g
10-491-4100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-491-4500G Supplier Sigma Aldrich Supplier No. 104914500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Dichloromethylvinylsilane reacts with dimethylsulphide borane to form organosilylborane, which further gets ammonolysized to form the polymer of the sibcn.dichloromethylvinylsilane was used as single molecular precursor for the deposition of α-sic thin films and α -sic nanowires by metalorganic chemical vapor deposition on bare si 100 and ni-coated si 100 substrates
  • Dichloromethylvinylsilane was used in the synthesis and characterization of tris(1-(dichloro(methyl)silyl)ethyl)borane,monomer

Specifications

Boiling Point 92°C (lit.)
CAS 124-70-9
Quantity 500 g
Molecular Formula C3H6Cl2Si
Linear Formula H2C=CHSi(CH3)Cl2
MDL Number MFCD00000492
Synonym Methylvinyldichlorosilane
Molecular Weight (g/mol) 141.07
Percent Purity 97%
Refractive Index n20/D 1.430 (literature)
Density 1.08 g/mL (at 25°C (literature))
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.