Learn More
Medchemexpress LLC HY-113950 1mg Medchemexpress, Dichlorophenyl-ABA CAS:18201-65-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1139501MG
Encompass_Preferred
Medchemexpress, HY-113950 1mg Dichlorophenyl-ABA CAS:18201-65-5 Dichlorophenyl-ABA is an inhibitor of transthyretin (TTR) amyloid fibril formation, inhibiting aggregate formation in more than 80% in TTR L55P-expressing cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9263