Learn More
Medchemexpress LLC Dichlorophenylindophenol sodium | 620-45-1 | MFCD00012176 | 98.0% | 5 G
SDP
Catalog No. 5000410140
Encompass_Preferred
DCIP sodium is a blue dye commonly used in various biochemical and biotechnological applications as an indicator of redox reactions. It possesses unique chemical properties that cause its color to change based on the oxidation state of the tested substance. It is frequently employed in enzyme assays, such as measuring succinate dehydrogenase activity, and in protein quantification methods like the Lowry assay.
  • Biochemical reagent.
  • Can be used as a biological material or organic compound for life science related research.
  • Blue dye that changes color according to the oxidation state of the substance being tested.
Catalog No. 50-004-10140 Supplier Medchemexpress LLC Supplier No. HYD00185G
DCIP sodium is a blue dye commonly used in various biochemical and biotechnological applications as an indicator of redox reactions. It possesses unique chemical properties that cause its color to change based on the oxidation state of the tested substance. It is frequently employed in enzyme assays, such as measuring succinate dehydrogenase activity, and in protein quantification methods like the Lowry assay.
  • Biochemical reagent.
  • Can be used as a biological material or organic compound for life science related research.
  • Blue dye that changes color according to the oxidation state of the substance being tested.