Learn More
Dichlorophenylphosphine, 97%, Thermo Scientific Chemicals
Description
Dichlorophenylphosphine and its derivatives are used as an intermediate to make plasticizers, antioxidants, stabilizers, catalysts and high pressure lubricant additives and in lacquer formulatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dichlorophenylphosphine and its derivatives are used as an intermediate to make plasticizers, antioxidants, stabilizers, catalysts and high pressure lubricant additives and in lacquer formulatings.
Solubility
Reacts with water.
Notes
Air & moisture Sensitive. Incompatible with bases, oxidizing agents, water/ moisture &air. Store under inert gas.
Specifications
Melting Point | -51°C |
Density | 1.319 |
Boiling Point | 99°C to 101°C (11 mmHg) |
Flash Point | 102°C (215°F) |
Odor | Pungent |
Refractive Index | 1.598 |
Quantity | 50 g |
UN Number | UN2798 |
Beilstein | 508189 |
Sensitivity | Air and moisture sensitive |
Show More |
Safety and Handling
Hazard Category | H250-H301-H314-H335 |
Hazard Statement | GHS H Statement H300-H314-H318 Fatal if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P210-P222-P260-P264b-P270-P271-P280-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
DOTInformation | Hazard Class: 4.2; Packaging Group: I |
EINECSNumber | 211-425-8 |
RTECSNumber | TB2478000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.