Learn More
Strem, An Ascensus Company CAS# 22594-69-0. 2g. Dichlorotricarbonylruthenium(II) dimer, min. 98%. MFCD00011528
SDP
Supplier:  Strem, An Ascensus Company 4404502G
Encompass
CAS# 22594-69-0. 2g. Dichlorotricarbonylruthenium(II) dimer, min. 98%. MFCD00011528. Molecular Weight: 512.02. Molecular Formula: [RuCl2(CO)3]2. Color/form: off-white xtl. Strem# 44-0450. http://www.strem.com/catalog/v/44-0450/
Catalog No. 50-901-11932