Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dichlorotris(1,10-phenanthroline)ruthenium(II) hydrate

Catalog No. 3437141G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-371-41G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-371-41G Supplier Sigma Aldrich Supplier No. 3437141G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The oxygen sensitivity and phototoxicity of [Ru(Phen)] in chicken embryo chorioallantoic membrane was investigated.Fluorescence quenching of dichloro(tris-1,10-phenanthroline) ruthenium (II) hydrate is is triggered by oxygen.1 (Ru(phen)3Cl2 may be used to imprint mesoporous molecular sieves (SBA-15).

Specifications

CAS 207802-45-7
Molecular Formula C36H24Cl2N6Ru -+ xH2O
Linear Formula C36H24Cl2N6Ru -+ xH2O
MDL Number MFCD00799695
Quantity 1 g
Synonym Tris(1,10-phenanthroline)ruthenium(II) chloride hydrate
Molecular Weight (g/mol) 712.59
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.