Learn More
Strem, An Ascensus Company DICHLOROTRIS(TRIPHENYLPHOSP-5G
SDP
Catalog No. 5090111934
Encompass
Dichlorotris(triphenylphosphine)ruthenium (II), 99%; Size: 5g; [15529-49-4]; RuCl2(P(C6H5)3)3; Mol Wt:958.85; Color & Form: black xtl.
Catalog No. 50-901-11934 Supplier Strem, An Ascensus Company Supplier No. 4405005G
Dichlorotris(triphenylphosphine)ruthenium (II), 99%; Size: 5g; [15529-49-4]; RuCl2(P(C6H5)3)3; Mol Wt:958.85; Color & Form: black xtl.