Learn More
Medchemexpress LLC Dichloroxylenol | 133-53-9 | MFCD00019981 | 95.4% | 191.06 g/mol | C8H8Cl2O | 50 G
SDP
Catalog No. 5000282725
Encompass_Preferred
Dichloroxylenol is a bactericidal preservative and deodorant supplied as a research reagent for laboratory and veterinary studies. It is commonly used in investigations of reproductive tract infections in livestock and is provided with documented analytical characterization for research use only.
  • Bactericidal preservative and deodorant
  • Used in research of reproductive tract infections in livestock
  • Supplied with documented purity for laboratory applications
  • Available in multiple pack sizes to suit experimental needs
  • Characterized by molecular formula and molecular weight for reproducibility
Catalog No. 50-002-82725 Supplier Medchemexpress LLC Supplier No. HYW14079950G
Add to Cart
Add to Cart
Dichloroxylenol is a bactericidal preservative and deodorant supplied as a research reagent for laboratory and veterinary studies. It is commonly used in investigations of reproductive tract infections in livestock and is provided with documented analytical characterization for research use only.
  • Bactericidal preservative and deodorant
  • Used in research of reproductive tract infections in livestock
  • Supplied with documented purity for laboratory applications
  • Available in multiple pack sizes to suit experimental needs
  • Characterized by molecular formula and molecular weight for reproducibility