Learn More
Cayman Chemical DIchlorphenamIde 50mg
SDP
Catalog No. 502994730
Encompass_Preferred
A carbonic anhydrase inhibitor (Kis = 1.20, 38, 50, and 50 nM for the human CA isoforms CAI, CAII, CAIX, and CAXII, respectively); lowers intraocular pressure in rabbits when applied topically in a 10% solution; rescues the potassium deficiency and prevents insulin-induced paralysis in a rat model of familial hypokalemic periodic paralysis when administered at a dose of 5.6 mg/kg per day for ten days
Catalog No. 50-299-4730 Supplier Cayman Chemical Supplier No. 2365850 MG
A carbonic anhydrase inhibitor (Kis = 1.20, 38, 50, and 50 nM for the human CA isoforms CAI, CAII, CAIX, and CAXII, respectively); lowers intraocular pressure in rabbits when applied topically in a 10% solution; rescues the potassium deficiency and prevents insulin-induced paralysis in a rat model of familial hypokalemic periodic paralysis when administered at a dose of 5.6 mg/kg per day for ten days