Learn More
Cayman Chemical Dichlorvos-d6, Purity: ≥99% deuterated forms (d1-d6), CAS: 203645-53-8, 5 mg
SDP
Catalog No. 502947908
Encompass_Preferred
An internal standard for the quantification of dichlorvos by GC- or LC-MS
Catalog No. 50-294-7908 Supplier Cayman Chemical Supplier No. 345175 MG
An internal standard for the quantification of dichlorvos by GC- or LC-MS