Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Dichotomous Identification Key, Macroinvertebrates
SDPSCI_ED

Catalog No. S81342 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
6/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S81342 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81342 Supplier LaMotte™ Supplier No. 5882DK6
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Spiral bound dichotomous key for identification of aquatic macroinvertebrates.

  • Identification to Order and some Families
  • Color-coded based on Pollution Tolerance level
  • Includes glossary, body part illutrations and size references
  • English and Spanish.

Specifications

Grade 3+
Quantity 6/Pk.
Product Type Laminated Dichotomous Key
For Use With (Application) Identification of Aquatic Macroinvertebrates
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.