Learn More
Medchemexpress LLC Diclazuril | 101831-37-2 | 99.7% | 500 MG
SDP
Catalog No. 5000405361
Encompass_Preferred
Diclazuril is a benzeneacetonitrile derivative and a potent, orally active anticoccidial agent. It is used for research into infectious and parasitic diseases such as coccidiosis, acute toxoplasmosis, and equine protozoal pyoencephalitis (EPM).
  • Potent and orally active anticoccidial agent
  • Used for research into infectious and parasitic diseases, including coccidiosis, acute toxoplasmosis, and equine protozoal pyoencephalitis (EPM)
  • Effective against *E. maxima* later in its life cycle
  • Reduces lesion scores and improves performance and health in challenged birds
  • Soluble in DMSO at 25 mg/mL (61.33 mM)
Catalog No. 50-004-05361 Supplier Medchemexpress LLC Supplier No. HYB0357500MG
Diclazuril is a benzeneacetonitrile derivative and a potent, orally active anticoccidial agent. It is used for research into infectious and parasitic diseases such as coccidiosis, acute toxoplasmosis, and equine protozoal pyoencephalitis (EPM).
  • Potent and orally active anticoccidial agent
  • Used for research into infectious and parasitic diseases, including coccidiosis, acute toxoplasmosis, and equine protozoal pyoencephalitis (EPM)
  • Effective against *E. maxima* later in its life cycle
  • Reduces lesion scores and improves performance and health in challenged birds
  • Soluble in DMSO at 25 mg/mL (61.33 mM)