Learn More
Apexbio Technology LLC Diclazuril 101831-37-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062279
Encompass_Preferred
Diclazuril is a benzeneacetonitrile derivative displaying anticoccidial activity by targeting developmental stages of parasites It inhibits both asexual replication and sexual gametogenesis phases of coccidia thus disrupting the parasite s proliferative and reproductive cycle In pharmacological studies Diclazuril exhibits inhibitory efficacy against various species of Eimeria parasites associated with coccidial infections in vitro and in vivo with reported IC50 values typically ranging from 0 01 to 2 M depending on parasite strain and assay conditions Additionally this compound serves as an investigative tool in experimental models studying acute Toxoplasma gondii infection pathogenesis parasite-host interactions and therapeutic intervention strategies as well as research on protozoal mechanisms in equine protozoal myeloencephalitis (EPM)
Catalog No. 50-000-62279 Supplier Apexbio Technology LLC Supplier No. B19275.1
Diclazuril is a benzeneacetonitrile derivative displaying anticoccidial activity by targeting developmental stages of parasites It inhibits both asexual replication and sexual gametogenesis phases of coccidia thus disrupting the parasite s proliferative and reproductive cycle In pharmacological studies Diclazuril exhibits inhibitory efficacy against various species of Eimeria parasites associated with coccidial infections in vitro and in vivo with reported IC50 values typically ranging from 0 01 to 2 M depending on parasite strain and assay conditions Additionally this compound serves as an investigative tool in experimental models studying acute Toxoplasma gondii infection pathogenesis parasite-host interactions and therapeutic intervention strategies as well as research on protozoal mechanisms in equine protozoal myeloencephalitis (EPM)