Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diclazuril-d4 | 1632495-80-7 | 97.8% | 1 MG
SDP

Catalog No. 5000406285
Encompass_Preferred

Diclazuril-d4 is a deuterium labeled Diclazuril, a potent and orally active anticoccidial agent. It is used in the research of infectious and parasitic diseases like coccidiosis, acute toxoplasmosis, and equine protozoal pyoencephalitis (EPM). It can also serve as a tracer and internal standard for quantitative analysis.

  • Deuterium labeled form of Diclazuril
  • Potent and orally active anticoccidial agent
  • Tracer and internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Molecular weight: 411.66
  • Formula: C17H5D4Cl3N4O2
  • Appearance: Solid, white to yellow
  • Purity: 97.8%

Catalog No. 50-004-06285 Supplier Medchemexpress LLC Supplier No. HYB0357S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diclazuril-d4 is a deuterium labeled Diclazuril, a potent and orally active anticoccidial agent. It is used in the research of infectious and parasitic diseases like coccidiosis, acute toxoplasmosis, and equine protozoal pyoencephalitis (EPM). It can also serve as a tracer and internal standard for quantitative analysis.

  • Deuterium labeled form of Diclazuril
  • Potent and orally active anticoccidial agent
  • Tracer and internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Molecular weight: 411.66
  • Formula: C17H5D4Cl3N4O2
  • Appearance: Solid, white to yellow
  • Purity: 97.8%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.