Learn More
Medchemexpress LLC Diclofenac | 15307-86-5 | 5 G
SDP
Catalog No. 5000375337
Encompass_Preferred
Diclofenac is a potent and nonselective anti-inflammatory agent that acts as a Cyclooxygenase (COX) inhibitor. It inhibits both human COX-1 and COX-2 enzymes, and has also been shown to induce apoptosis of neural stem cells (NSCs) by activating the caspase cascade. It is intended for research use only.
  • Potent and nonselective anti-inflammatory agent
  • Acts as a COX inhibitor
  • Induces apoptosis of neural stem cells (NSCs)
  • For research use only
Catalog No. 50-003-75337 Supplier Medchemexpress LLC Supplier No. HY150365G
Diclofenac is a potent and nonselective anti-inflammatory agent that acts as a Cyclooxygenase (COX) inhibitor. It inhibits both human COX-1 and COX-2 enzymes, and has also been shown to induce apoptosis of neural stem cells (NSCs) by activating the caspase cascade. It is intended for research use only.
  • Potent and nonselective anti-inflammatory agent
  • Acts as a COX inhibitor
  • Induces apoptosis of neural stem cells (NSCs)
  • For research use only