Learn More
Apexbio Technology LLC Diclofenac 15307-86-5 5g
SDP
Catalog No. 502240675
Encompass_Preferred
Diclofenac (CAS 15307-86-5) is a phenylacetic acid derivative classified as a non-steroidal anti-inflammatory drug (NSAID) It exerts its biological activity by inhibiting cyclooxygenase (COX)-1 and COX-2 enzymes thereby suppressing prostaglandin synthesis that mediates inflammation and pain signaling Structurally the incorporation of two ortho chlorine atoms on the phenyl ring increases molecular rigidity which has been associated with enhanced potency Diclofenac is widely used in research to study inflammatory pathways pain mechanisms and as a reference compound in the evaluation of cyclooxygenase inhibition and potential gastrointestinal protective strategies in combination therapies
This item is not returnable. View return policy
Catalog No. 50-224-0675 Supplier Apexbio Technology LLC Supplier No. B35055000
Diclofenac (CAS 15307-86-5) is a phenylacetic acid derivative classified as a non-steroidal anti-inflammatory drug (NSAID) It exerts its biological activity by inhibiting cyclooxygenase (COX)-1 and COX-2 enzymes thereby suppressing prostaglandin synthesis that mediates inflammation and pain signaling Structurally the incorporation of two ortho chlorine atoms on the phenyl ring increases molecular rigidity which has been associated with enhanced potency Diclofenac is widely used in research to study inflammatory pathways pain mechanisms and as a reference compound in the evaluation of cyclooxygenase inhibition and potential gastrointestinal protective strategies in combination therapies