Learn More
AdipoGen Diclofenac acid
SDP
Supplier:  AdipoGen CDXD0147G100
Encompass_Preferred
Chemical. CAS 15307-86-5. Formula C14H11Cl2NO2. MW 296.15. Synthetic Nonsteroidal anti-inflammatory drug NSAID taken or applied to reduce inflammation and as an analgesic reducing pain. The primary mechanism responsible for its anti-inflammatory, antipyretic, and analgesic action is thought to be inhibition of prostaglandin synthesis by inhibition of cyclooxygenase COX. It also appears to exhibit bacteriostatic activity by inhibiting bacterial DNA synthesis. Can be used as a reference compound.
Catalog No. 50-256-1213