Learn More
Cayman Chemical Diclofenac Acyl-β-D-Glucuronide, 64118-81-6, 1 mg
SDP
Supplier:  Cayman Chemical 224551
Encompass_Preferred
A metabolite of diclofenac; acutely toxic in vitro; may play a role in the hepatotoxicity of diclofenac in vivo; binds OATs (Km = 60.2, 8.6, 112, and 103.9 µM for OAT1-4, respectively); may be back-transformed to diclofenac in wastewater,
Catalog No. NC1506720