Learn More
Cayman Chemical Diclofenac-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 153466-65-0, 1 mg
SDP
Catalog No. 502984894
Encompass_Preferred
An internal standard for the quantification of diclofenac by GC- or LC-MS
Catalog No. 50-298-4894 Supplier Cayman Chemical Supplier No. 190721 MG
An internal standard for the quantification of diclofenac by GC- or LC-MS