Learn More
Cayman Chemical DIclofenac-d4 500ug
SDP
Catalog No. 502981418
Encompass_Preferred
An internal standard for the quantification of diclofenac by GC- or LC-MS
Catalog No. 50-298-1418 Supplier Cayman Chemical Supplier No. 19072500 UG
An internal standard for the quantification of diclofenac by GC- or LC-MS