Learn More
Medchemexpress LLC Diclofenac-d4 sodium | 154523-54-3 | C14H6D4Cl2NNaO2 | 1 MG
SDP
Catalog No. 5000375839
Encompass_Preferred
Diclofenac-d4 (sodium) is the deuterium labeled Diclofenac sodium. It is a potent and nonselective anti-inflammatory agent that acts as a COX inhibitor. It also induces apoptosis of neural stem cells (NSCs) via the activation of the caspase cascade. This product is for research use only and not for sale to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Acts as a potent and nonselective anti-inflammatory agent
  • Inhibits COX enzymes
  • Induces apoptosis of neural stem cells (NSCs)
Catalog No. 50-003-75839 Supplier Medchemexpress LLC Supplier No. HY15037S11MG
Diclofenac-d4 (sodium) is the deuterium labeled Diclofenac sodium. It is a potent and nonselective anti-inflammatory agent that acts as a COX inhibitor. It also induces apoptosis of neural stem cells (NSCs) via the activation of the caspase cascade. This product is for research use only and not for sale to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Acts as a potent and nonselective anti-inflammatory agent
  • Inhibits COX enzymes
  • Induces apoptosis of neural stem cells (NSCs)