Learn More
Apexbio Technology LLC Diclofenac Diethylamine 78213-16-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092725
Encompass_Preferred
Diclofenac diethylamine (CAS 78213-16-8) is a small molecule classified as a nonsteroidal anti-inflammatory drug (NSAID) It exerts its pharmacological effects primarily through the inhibition of cyclooxygenase (COX) enzymes resulting in reduced synthesis of prostaglandins involved in inflammation and pain signaling Diclofenac diethylamine is frequently utilized in preclinical and clinical research to investigate inflammatory pathways and nociception as well as to model therapeutic interventions for musculoskeletal and rheumatological disorders Its well-characterized mechanism supports its use for studies focusing on analgesic and anti-inflammatory pharmacodynamics
Catalog No. 50-000-92725 Supplier Apexbio Technology LLC Supplier No. B19285.1
Diclofenac diethylamine (CAS 78213-16-8) is a small molecule classified as a nonsteroidal anti-inflammatory drug (NSAID) It exerts its pharmacological effects primarily through the inhibition of cyclooxygenase (COX) enzymes resulting in reduced synthesis of prostaglandins involved in inflammation and pain signaling Diclofenac diethylamine is frequently utilized in preclinical and clinical research to investigate inflammatory pathways and nociception as well as to model therapeutic interventions for musculoskeletal and rheumatological disorders Its well-characterized mechanism supports its use for studies focusing on analgesic and anti-inflammatory pharmacodynamics