Learn More
Medchemexpress LLC Diclofenac epolamine | 119623-66-4 | 100.0% | 411.32 | 50 MG
SDP
Catalog No. 5000345233
Encompass_Preferred
Diclofenac epolamine is a non-steroidal anti-inflammatory drug (NSAID) used for relieving arthritis pain, acute pain, osteoarthritis, and actinic keratosis. It demonstrates good skin absorption characteristics without local adverse reactions or allergies.
  • Relieves arthritis pain, acute pain, osteoarthritis, and actinic keratosis.
  • Demonstrates good skin absorption characteristics.
  • Does not cause local adverse reactions or allergies.
  • Effective in treating inflammatory conditions.
  • Increases partitioning into the skin.
  • Enhances and sustains drug transport through the skin.
Catalog No. 50-003-45233 Supplier Medchemexpress LLC Supplier No. HY10954710MM/1ML
Diclofenac epolamine is a non-steroidal anti-inflammatory drug (NSAID) used for relieving arthritis pain, acute pain, osteoarthritis, and actinic keratosis. It demonstrates good skin absorption characteristics without local adverse reactions or allergies.
  • Relieves arthritis pain, acute pain, osteoarthritis, and actinic keratosis.
  • Demonstrates good skin absorption characteristics.
  • Does not cause local adverse reactions or allergies.
  • Effective in treating inflammatory conditions.
  • Increases partitioning into the skin.
  • Enhances and sustains drug transport through the skin.