Learn More
Medchemexpress LLC Diclofenac ethyl ester | 15307-77-4 | 99.3% | 100 MG
SDP
Catalog No. 5000339227
Encompass_Preferred
Diclofenac ethyl ester is a prodrug of Diclofenac, designed for enhanced delivery. It is encapsulated in polymer micelles in vitro and released with prolonged circulation time and reduced drug concentrations in the heart and kidneys in vivo.
  • Prodrug of Diclofenac
  • Encapsulated in polymer micelles
  • Prolonged circulation time
  • Reduced drug concentrations in heart and kidneys
Catalog No. 50-003-39227 Supplier Medchemexpress LLC Supplier No. HY127125100MG
Diclofenac ethyl ester is a prodrug of Diclofenac, designed for enhanced delivery. It is encapsulated in polymer micelles in vitro and released with prolonged circulation time and reduced drug concentrations in the heart and kidneys in vivo.
  • Prodrug of Diclofenac
  • Encapsulated in polymer micelles
  • Prolonged circulation time
  • Reduced drug concentrations in heart and kidneys