Learn More
Medchemexpress LLC Diclofenac ethyl ester | 15307-77-4 | 99.3% | 50 MG
SDP
Catalog No. 5000341302
Encompass_Preferred
Diclofenac ethyl ester is the prodrug of Diclofenac, encapsulated in polymer micelles in vitro. It is released with prolonged circulation time and reduced drug concentrations in the heart and kidneys in vivo.
  • Prodrug of Diclofenac
  • Encapsulated in polymer micelles
  • Designed for prolonged circulation time
  • Aids in reduced drug concentrations in heart and kidneys
  • Targets COX
  • Relevant to immunology and inflammation pathways
  • Soluble in DMSO at 50 mg/mL (requires ultrasonic)
  • Soluble in 10% DMSO and 90% corn oil for in vivo applications (≥ 2.5 mg/mL)
Catalog No. 50-003-41302 Supplier Medchemexpress LLC Supplier No. HY12712550MG
Diclofenac ethyl ester is the prodrug of Diclofenac, encapsulated in polymer micelles in vitro. It is released with prolonged circulation time and reduced drug concentrations in the heart and kidneys in vivo.
  • Prodrug of Diclofenac
  • Encapsulated in polymer micelles
  • Designed for prolonged circulation time
  • Aids in reduced drug concentrations in heart and kidneys
  • Targets COX
  • Relevant to immunology and inflammation pathways
  • Soluble in DMSO at 50 mg/mL (requires ultrasonic)
  • Soluble in 10% DMSO and 90% corn oil for in vivo applications (≥ 2.5 mg/mL)