Learn More
Cayman Chemical DIclofenac ethyl ester 25mg
SDP
Catalog No. 502964193
Encompass_Preferred
An esterified form of diclofenac; has been used as a precursor in the synthesis of diclofenac derivatives with anti-inflammatory and analgesic activity and decreased ulcerogenicity
Catalog No. 50-296-4193 Supplier Cayman Chemical Supplier No. 2210525 MG
An esterified form of diclofenac; has been used as a precursor in the synthesis of diclofenac derivatives with anti-inflammatory and analgesic activity and decreased ulcerogenicity