Learn More
AdipoGen Diclofenac Impurity C
SDP
Supplier:  AdipoGen CDXD1227M050
Encompass_Preferred
Chemical. CAS 27204-57-5. Formula C13H11Cl2NO. MW 268.14. Diclofenac Impurity C is a degradation product of and a regulatory impurity in the drug Diclofenac which is a non-steroidal anti-inflammatory drug NSAID that is clinically used to treat inflammation associated with arthritis and gout as well as other pain or inflammatory disorders. This product can be used as a working standard or secondary analytical reference standard. It can also be used as an organic building block and intermediate for synthesis.
Catalog No. 50-256-1009