Learn More
Apexbio Technology LLC Diclofenac Potassium 15307-81-0 5g
SDP
Catalog No. 5000094340
Encompass_Preferred
Diclofenac potassium (CAS 15307-81-0) is a synthetic small molecule belonging to the nonsteroidal anti-inflammatory drug (NSAID) class It functions primarily by inhibiting cyclooxygenase (COX) enzymes thereby reducing the synthesis of prostaglandins involved in inflammation and nociception Through this mechanism diclofenac potassium attenuates inflammatory responses and mediates analgesic effects It is frequently utilized in biomedical research to study inflammatory pathways investigate mechanisms of pain modulation and evaluate anti-inflammatory interventions in cellular and preclinical models
Catalog No. 50-000-94340 Supplier Apexbio Technology LLC Supplier No. B19295000
Diclofenac potassium (CAS 15307-81-0) is a synthetic small molecule belonging to the nonsteroidal anti-inflammatory drug (NSAID) class It functions primarily by inhibiting cyclooxygenase (COX) enzymes thereby reducing the synthesis of prostaglandins involved in inflammation and nociception Through this mechanism diclofenac potassium attenuates inflammatory responses and mediates analgesic effects It is frequently utilized in biomedical research to study inflammatory pathways investigate mechanisms of pain modulation and evaluate anti-inflammatory interventions in cellular and preclinical models