Learn More
Medchemexpress LLC Diclofenac potassium | 15307-81-0 | C14H10Cl2KNO2 | 500 MG
SDP
Catalog No. 5000375123
Encompass_Preferred
Diclofenac potassium is a potent and nonselective anti-inflammatory agent that acts as a COX inhibitor. It is harmful if swallowed and is toxic to aquatic life with long-lasting effects. Use only outdoors or in a well-ventilated area, and wear appropriate personal protective equipment.
  • Potent and nonselective anti-inflammatory agent
  • Acts as a COX inhibitor
  • Used for research and analytical applications
  • Available as a solid for various laboratory needs
Catalog No. 50-003-75123 Supplier Medchemexpress LLC Supplier No. HY15038500MG
Add to Cart
Add to Cart
Diclofenac potassium is a potent and nonselective anti-inflammatory agent that acts as a COX inhibitor. It is harmful if swallowed and is toxic to aquatic life with long-lasting effects. Use only outdoors or in a well-ventilated area, and wear appropriate personal protective equipment.
  • Potent and nonselective anti-inflammatory agent
  • Acts as a COX inhibitor
  • Used for research and analytical applications
  • Available as a solid for various laboratory needs