Learn More
Medchemexpress LLC Diclofenac potassium | 15307-81-0 | MFCD00082254 | C14H10Cl2KNO2 | 5 G
SDP
Catalog No. 5000375173
Encompass_Preferred
Diclofenac potassium is a potent and nonselective anti-inflammatory agent that acts as a COX inhibitor. It has IC50s of 4 nM for human COX-1 and 1.3 nM for human COX-2 in CHO cells. It also induces apoptosis of neural stem cells (NSCs) through the activation of the caspase cascade.
  • Potent and nonselective anti-inflammatory agent
  • Acts as a COX inhibitor
  • Induces apoptosis of neural stem cells (NSCs) by activating the caspase cascade
  • Blocks COX-1 mediated prostanoid production
  • For research use only
Catalog No. 50-003-75173 Supplier Medchemexpress LLC Supplier No. HY150385G
Add to Cart
Add to Cart
Diclofenac potassium is a potent and nonselective anti-inflammatory agent that acts as a COX inhibitor. It has IC50s of 4 nM for human COX-1 and 1.3 nM for human COX-2 in CHO cells. It also induces apoptosis of neural stem cells (NSCs) through the activation of the caspase cascade.
  • Potent and nonselective anti-inflammatory agent
  • Acts as a COX inhibitor
  • Induces apoptosis of neural stem cells (NSCs) by activating the caspase cascade
  • Blocks COX-1 mediated prostanoid production
  • For research use only