Learn More
Apexbio Technology LLC Diclofenac Sodium 15307-79-6 5g
SDP
Catalog No. 502240315
Encompass_Preferred
Diclofenac Sodium (CAS 15307-79-6) is a non-steroidal anti-inflammatory compound that inhibits cyclooxygenase (COX) enzymes It acts as a non-selective COX inhibitor demonstrating IC50 values of 60 nM for ovine COX-1 and 220 nM for COX-2 By suppressing prostaglandin synthesis Diclofenac Sodium exhibits anti-inflammatory antipyretic and analgesic effects It is widely utilized in biomedical research to investigate inflammatory pathways pain mechanisms and the pharmacological modulation of COX activity
Catalog No. 50-224-0315 Supplier Apexbio Technology LLC Supplier No. B14575000
Diclofenac Sodium (CAS 15307-79-6) is a non-steroidal anti-inflammatory compound that inhibits cyclooxygenase (COX) enzymes It acts as a non-selective COX inhibitor demonstrating IC50 values of 60 nM for ovine COX-1 and 220 nM for COX-2 By suppressing prostaglandin synthesis Diclofenac Sodium exhibits anti-inflammatory antipyretic and analgesic effects It is widely utilized in biomedical research to investigate inflammatory pathways pain mechanisms and the pharmacological modulation of COX activity