Learn More
Selleck Chemical LLC Diclofenac Sodium 50mg 15307-79-6 GP 45840
SDP
Catalog No. 507572
Encompass_Preferred
Diclofenac Sodium (GP 45840) is a non-selective COX inhibitor with IC50 of 0.5 ?g/ml and 0.5 ?g/ml for COX-1 and -2 in intact cells, respectively, used as a nonsteroidal anti-inflammatory drug (NSAID) to relieve pain and reduce swelling in flammation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-757-2 Supplier Selleck Chemical LLC Supplier No. S190350MG
Diclofenac Sodium (GP 45840) is a non-selective COX inhibitor with IC50 of 0.5 ?g/ml and 0.5 ?g/ml for COX-1 and -2 in intact cells, respectively, used as a nonsteroidal anti-inflammatory drug (NSAID) to relieve pain and reduce swelling in flammation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.