Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diclofenac, sodium salt, 100.38%, MP Biomedicals™

Catalog No. ICN15766010 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15766010 10 g
ICN15766080 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15766010 Supplier MP Biomedicals, Inc Supplier No. 0215766010
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15307-79-6
Molecular Formula C14H10Cl2NNaO2
Molecular Weight (g/mol) 318.129
InChI Key KPHWPUGNDIVLNH-UHFFFAOYSA-M
Synonym diclofenac sodium, sodium diclofenac, diclofenac sodium salt, voltaren, solaraze, anthraxiton, prophenatin, allvoran, batafil, delimon
PubChem CID 5018304
ChEBI CHEBI:4509
IUPAC Name sodium;2-[2-(2,6-dichloroanilino)phenyl]acetate
SMILES C1=CC=C(C(=C1)CC(=O)[O-])NC2=C(C=CC=C2Cl)Cl.[Na+]

Specifications

Color White
Density 0.55 to 0.7g/mL
Assay Percent Range 1.0038
Quantity 10 g
Formula Weight 318.1
Physical Form Powder
Chemical Name or Material Diclofenac, sodium salt
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.