Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diclofenac sodium salt, Analytical Standard, ≥98.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111019875 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9875 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9875 Supplier MilliporeSigma Supelco Supplier No. 93484100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 15307-79-6
Physical Form Neat
Linear Formula C14H10Cl2NNaO2
MDL Number MFCD00082251
Synonym 2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid sodium salt
Quantity 100 mg
Formula Weight 318.13
Percent Purity ≥98.5% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.