Learn More
Abcam Diclofenac sodium salt, cyclo-o x ygenase inhibitor, 1G
Catalog No. 502707413
Encompass_Preferred
MW 318.13 g/mol, Purity >99%. Potent non-selective COX inhibitor (IC₅₀ values are 75 and 29 nM at COX-1 and COX-2, respectively). Potent inhibitor of phospholipase A₂ in vitro and in vivo. Activates the nitric oxide-cyclic GMP-potassium channel pathway. Shows anti-inflammatory, antipyretic and analgesic effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7413 Supplier Abcam Supplier No. AB1206211G
MW 318.13 g/mol, Purity >99%. Potent non-selective COX inhibitor (IC₅₀ values are 75 and 29 nM at COX-1 and COX-2, respectively). Potent inhibitor of phospholipase A₂ in vitro and in vivo. Activates the nitric oxide-cyclic GMP-potassium channel pathway. Shows anti-inflammatory, antipyretic and analgesic effects.
The product is subject to the following: Abcam Restricted Use Statement