Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diclofenac Sodium, USP, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18602176 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-176 25 g Amber Glass Bottle
18-602-177 100 g Amber Glass Bottle
18-602-178 500 g Amber Glass Bottle
18-602-181 1 kg Amber Glass Bottle
18-602-179 5 kg Poly Pail
18-602-180 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-602-176 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D128725GM
Add to Cart
Add to Cart

D1287, 15307-79-6, C14H10Cl2NNaO2

Spectrum™ Chemical Diclofenac Sodium, USP is a nonsteroidal anti-inflammatory drug used to reduce inflammation and is as an analgesic. All Spectrum Chemical USP products are manufactured, packa ged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 15307-79-6
Molecular Formula C14H10Cl2NNaO2
Molecular Weight (g/mol) 318.13
InChI Key KPHWPUGNDIVLNH-UHFFFAOYSA-M
IUPAC Name sodium 2-{2-[(2,6-dichlorophenyl)amino]phenyl}acetate
SMILES [Na+].[O-]C(=O)CC1=CC=CC=C1NC1=C(Cl)C=CC=C1Cl

Specifications

CAS Max % 100%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 25 g
UN Number UN2811
Percent Purity 99 to 101%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.