Learn More
Cayman Chemical DIclofenacsodIum salt 25g
SDP
Catalog No. 502896709
Encompass_Preferred
An NSAID and a COX inhibitor (IC50s = 0.9-2.7 and 1.5-20 µM, for human COX-1 and COX-2, respectively); inhibits A23187-induced release of arachidonic acid in isolated rat peritoneal neutrophils and macrophages (IC50s = 60 and 10 µM, respectively); inhibits carrageenan-induced paw edema in rats
Catalog No. 50-289-6709 Supplier Cayman Chemical Supplier No. 7068025 G
An NSAID and a COX inhibitor (IC50s = 0.9-2.7 and 1.5-20 µM, for human COX-1 and COX-2, respectively); inhibits A23187-induced release of arachidonic acid in isolated rat peritoneal neutrophils and macrophages (IC50s = 60 and 10 µM, respectively); inhibits carrageenan-induced paw edema in rats