Learn More
Medchemexpress LLC Diclofensine (hydrochloride) | 34041-84-4 | 99.79% | 358.69 | 1 ML
SDP
Catalog No. 5000387975
Encompass_Preferred
Diclofensine hydrochloride is an orally active neuronal monoamine uptake inhibitor. It blocks the uptake of dopamine, noradrenaline, and serotonin, and can be used in the research of moderate to severe depression.
  • Orally active neuronal monoamine uptake inhibitor.
  • Blocks dopamine, noradrenaline, and serotonin uptake.
  • Potentiates norepinephrine-induced hypertension.
  • Attenuates Tyramine-induced hypertension.
  • Produces psychostimulant and mood-elevating effects.
  • Can be used in research of moderate to severe depression.
Catalog No. 50-003-87975 Supplier Medchemexpress LLC Supplier No. HY1861010MM/1ML
Diclofensine hydrochloride is an orally active neuronal monoamine uptake inhibitor. It blocks the uptake of dopamine, noradrenaline, and serotonin, and can be used in the research of moderate to severe depression.
  • Orally active neuronal monoamine uptake inhibitor.
  • Blocks dopamine, noradrenaline, and serotonin uptake.
  • Potentiates norepinephrine-induced hypertension.
  • Attenuates Tyramine-induced hypertension.
  • Produces psychostimulant and mood-elevating effects.
  • Can be used in research of moderate to severe depression.