Learn More
Medchemexpress LLC Diclofensine hydrochloride | 34041-84-4 | MFCD00865459 | 99.8% | 358.69 g/mol | C17H18Cl3NO | 10 MG
SDP
Catalog No. 5000295006
Encompass_Preferred
Diclofensine hydrochloride is a research reagent that potently inhibits monoamine reuptake and is used to study dopaminergic and other monoaminergic transporter function in vitro. The hydrochloride salt is supplied as a high-purity solid suitable for biochemical, pharmacological, and transporter assays.
  • Potent inhibitor of dopamine, noradrenaline, and serotonin uptake with low-nanomolar IC50 values.
  • Suitable for dopamine transporter and reuptake mechanism studies.
  • Supplied as a hydrochloride salt in solid form for consistent handling.
  • High purity (~99.8%) appropriate for biochemical assays.
  • Available in small pack sizes for laboratory-scale experiments.
  • Recommended storage sealed and away from moisture; in solvent: -80 °C (6 months) or -20 °C (1 month).
Catalog No. 50-002-95006 Supplier Medchemexpress LLC Supplier No. HY1861010MG
Diclofensine hydrochloride is a research reagent that potently inhibits monoamine reuptake and is used to study dopaminergic and other monoaminergic transporter function in vitro. The hydrochloride salt is supplied as a high-purity solid suitable for biochemical, pharmacological, and transporter assays.
  • Potent inhibitor of dopamine, noradrenaline, and serotonin uptake with low-nanomolar IC50 values.
  • Suitable for dopamine transporter and reuptake mechanism studies.
  • Supplied as a hydrochloride salt in solid form for consistent handling.
  • High purity (~99.8%) appropriate for biochemical assays.
  • Available in small pack sizes for laboratory-scale experiments.
  • Recommended storage sealed and away from moisture; in solvent: -80 °C (6 months) or -20 °C (1 month).