Learn More
Cayman Chemical Diclofensine (hydrochloride), Purity: ≥98%, CAS: 34041-84-4, 10 mg
SDP
Catalog No. 502971865
Encompass_Preferred
A potent inhibitor of monoamine reuptake, blocking the uptake of dopamine, noradrenaline, and serotonin by rat brain synaptosomes with IC50 values of 0.74, 2.3, and 3.7 nM, respectively; intended for research and forensic applications
Catalog No. 50-297-1865 Supplier Cayman Chemical Supplier No. 1535510 MG
A potent inhibitor of monoamine reuptake, blocking the uptake of dopamine, noradrenaline, and serotonin by rat brain synaptosomes with IC50 values of 0.74, 2.3, and 3.7 nM, respectively; intended for research and forensic applications