Learn More
Cayman Chemical Diclofensine (hydrochloride), Purity: ≥98%, CAS: 34041-84-4, 50 mg
SDP
Catalog No. 502969112
Encompass_Preferred
A potent inhibitor of monoamine reuptake, blocking the uptake of dopamine, noradrenaline, and serotonin by rat brain synaptosomes with IC50 values of 0.74, 2.3, and 3.7 nM, respectively; intended for research and forensic applications
Catalog No. 50-296-9112 Supplier Cayman Chemical Supplier No. 1535550 MG
A potent inhibitor of monoamine reuptake, blocking the uptake of dopamine, noradrenaline, and serotonin by rat brain synaptosomes with IC50 values of 0.74, 2.3, and 3.7 nM, respectively; intended for research and forensic applications