Learn More
AdipoGen Diclofop
SDP
Supplier:  AdipoGen CDXD0314M050
Encompass_Preferred
Chemical. CAS 40843-25-2. Formula C15H12Cl2O4. MW 327.2. Synthetic. Herbicide. Peroxisome proliferator in rodents with carcinogenic activity. Compound can be used as analytical reference material.
Catalog No. 50-256-1126