Learn More
Crescent Chemical Co Inc DICLOFOP (FREE ACID) 100 MG
SDP
Catalog No. 5073444
Encompass_Preferred
Diclofop (free acid) Insecticide (Metabolite neat) CAS 40843-25-2 Molecular Weight - 327.1594 Storage - RT Catalog Num - DRE-C12539000 pesticide
Catalog No. 50-734-44 Supplier Crescent Chemical Co Inc Supplier No. PSC12539000
Diclofop (free acid) Insecticide (Metabolite neat) CAS 40843-25-2 Molecular Weight - 327.1594 Storage - RT Catalog Num - DRE-C12539000 pesticide