Learn More
Medchemexpress LLC Diclosulam | 145701-21-9 | 98.2% | 500 MG
SDP
Catalog No. 5000341949
Encompass_Preferred
Diclosulam is a herbicide, also known by its synonym XDE 564. It is characterized by a Ki value of <32 nM and exhibits MIC and MIC50 values against CBS10913 and CBS12373 strains, ranging from 6.25 to 12.5 μM and 1.40 to 3.01 μM, respectively. Its applications extend to targeting acetohydroxyacid synthase.
  • Herbicide activity.
  • Potent inhibition of drug-resistant *Candida auris* growth.
  • Targets acetohydroxyacid synthase.
  • Available in solid appearance.
  • Purity of 98.20%.
Catalog No. 50-003-41949 Supplier Medchemexpress LLC Supplier No. HY12852500MG
Diclosulam is a herbicide, also known by its synonym XDE 564. It is characterized by a Ki value of <32 nM and exhibits MIC and MIC50 values against CBS10913 and CBS12373 strains, ranging from 6.25 to 12.5 μM and 1.40 to 3.01 μM, respectively. Its applications extend to targeting acetohydroxyacid synthase.
  • Herbicide activity.
  • Potent inhibition of drug-resistant *Candida auris* growth.
  • Targets acetohydroxyacid synthase.
  • Available in solid appearance.
  • Purity of 98.20%.