Learn More
Cayman Chemical Dicloxacillin (sodium salt hydrate), Purity: ≥98%, CAS: 13412-64-1, 1 g
SDP
Catalog No. 502964499
Encompass_Preferred
A semisynthetic penicillin antibiotic; has activity against Gram-positive bacteria (MICs = 0.02-0.24 μg/ml for S. pyogenes, S. aureus, and D. pneumoniae); protective against S. pyogenes, S. aureus, and D. pneumoniae infections in mice when administered subcutaneously or orally (PD50s = 12-185 and 41-280 mg/kg, respectively)
Catalog No. 50-296-4499 Supplier Cayman Chemical Supplier No. 237701 G
A semisynthetic penicillin antibiotic; has activity against Gram-positive bacteria (MICs = 0.02-0.24 μg/ml for S. pyogenes, S. aureus, and D. pneumoniae); protective against S. pyogenes, S. aureus, and D. pneumoniae infections in mice when administered subcutaneously or orally (PD50s = 12-185 and 41-280 mg/kg, respectively)