Learn More
Research Products International Corp Dicloxacillin Sodum Slt 100 MG
SDP
Catalog No. 502230524
Encompass_Preferred
Dicloxacillin sodium salt is a narrow-spectrum antibiotic in the penicillin class, specifically belonging to the beta-lactamase-resistant penicillins.
As a beta-lactam antibiotic, dicloxacillin works by inhibiting the synthesis of bacterial cell walls, leading to the destruction of the bacterial cell. 
Catalog No. 50-223-0524 Supplier Research Products International Corp Supplier No. D821500.1
Dicloxacillin sodium salt is a narrow-spectrum antibiotic in the penicillin class, specifically belonging to the beta-lactamase-resistant penicillins.
As a beta-lactam antibiotic, dicloxacillin works by inhibiting the synthesis of bacterial cell walls, leading to the destruction of the bacterial cell. 