Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dicloxacillin sodium | 343-55-5 | 98.0% | 25 MG
SDP

Catalog No. 5000414216
Encompass_Preferred

Dicloxacillin sodium is a β-lactam antibiotic belonging to the penicillin family. It is effective against Gram-positive bacteria and β-lactamase-producing organisms, such as Staphylococcus aureus.

  • Effective against Gram-positive bacteria
  • Active against β-lactamase-producing organisms
  • Purity of 98.0%
  • CAS No. 343-55-5
  • Molecular Weight: 492.31
  • Formula: C19H16Cl2N3NaO5S
  • Appearance: Solid
  • Color: White to off-white

Catalog No. 50-004-14216 Supplier Medchemexpress LLC Supplier No. 25MGHYB1459
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dicloxacillin sodium is a β-lactam antibiotic belonging to the penicillin family. It is effective against Gram-positive bacteria and β-lactamase-producing organisms, such as Staphylococcus aureus.

  • Effective against Gram-positive bacteria
  • Active against β-lactamase-producing organisms
  • Purity of 98.0%
  • CAS No. 343-55-5
  • Molecular Weight: 492.31
  • Formula: C19H16Cl2N3NaO5S
  • Appearance: Solid
  • Color: White to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.