Learn More
Selleck Chemical LLC Dicloxacillin Sodium hydrate
SDP
Catalog No. 5000004096
Encompass_Preferred
Dicloxacillin Sodium hydrate (DLX Dycill Dynapen Pathocil) a narrow-spectrum -Lactam antibiotic of the penicillin class induces drug-metabolizing CYP enzymes to a clinically relevant extent
Catalog No. 50-000-04096 Supplier Selleck Chemical LLC Supplier No. S596925MG
Dicloxacillin Sodium hydrate (DLX Dycill Dynapen Pathocil) a narrow-spectrum -Lactam antibiotic of the penicillin class induces drug-metabolizing CYP enzymes to a clinically relevant extent