Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dicloxacillin (Sodium hydrate) | 13412-64-1 | 98.9% | 1 ML
SDP

Catalog No. 5000408875
Encompass_Preferred

Dicloxacillin (Sodium hydrate) is a chemical compound with CAS Number 13412-64-1, molecular formula C19H18Cl2N3NaO6S, and molecular weight 510.32. It appears as a white to off-white solid with a purity of 98.94% (LCMS). This product is intended for laboratory chemical use and research purposes only, and is stable under recommended storage conditions.

  • Appears as a white to off-white solid
  • Has a purity of 98.94% (LCMS)
  • For research use only
  • Stable under recommended storage conditions
  • Store at 4°C, sealed, away from moisture
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-08875 Supplier Medchemexpress LLC Supplier No. HYB097710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dicloxacillin (Sodium hydrate) is a chemical compound with CAS Number 13412-64-1, molecular formula C19H18Cl2N3NaO6S, and molecular weight 510.32. It appears as a white to off-white solid with a purity of 98.94% (LCMS). This product is intended for laboratory chemical use and research purposes only, and is stable under recommended storage conditions.

  • Appears as a white to off-white solid
  • Has a purity of 98.94% (LCMS)
  • For research use only
  • Stable under recommended storage conditions
  • Store at 4°C, sealed, away from moisture
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.