Learn More
Medchemexpress LLC Dicloxacillin Sodium hydrate | 13412-64-1 | 98.9% | 5 MG
SDP
Catalog No. 5000408047
Encompass_Preferred
Dicloxacillin Sodium hydrate is a narrow-spectrum β-Lactam antibiotic of the penicillin class. It is used to treat infections caused by susceptible Gram-positive bacteria and is active against beta-lactamase-producing organisms such as Staphylococcus aureus.
  • Narrow-spectrum β-Lactam antibiotic of the penicillin class.
  • Used to treat infections caused by susceptible Gram-positive bacteria.
  • Active against beta-lactamase-producing organisms like Staphylococcus aureus.
  • Exhibits EC50 values of 0.06 and 0.50 mg/L in ATCC 25923 and E19977, respectively.
  • Exhibits MIC values of 0.125 and 0.5 mg/L in ATCC 25923 and E19977 with pH 7.4, respectively.
  • Demonstrates therapeutic activity in a murine peritonitis-sepsis model, with all mice surviving at a dosage of 125 mg/kg via IV injection.
Catalog No. 50-004-08047 Supplier Medchemexpress LLC Supplier No. HYB09775MG
Dicloxacillin Sodium hydrate is a narrow-spectrum β-Lactam antibiotic of the penicillin class. It is used to treat infections caused by susceptible Gram-positive bacteria and is active against beta-lactamase-producing organisms such as Staphylococcus aureus.
  • Narrow-spectrum β-Lactam antibiotic of the penicillin class.
  • Used to treat infections caused by susceptible Gram-positive bacteria.
  • Active against beta-lactamase-producing organisms like Staphylococcus aureus.
  • Exhibits EC50 values of 0.06 and 0.50 mg/L in ATCC 25923 and E19977, respectively.
  • Exhibits MIC values of 0.125 and 0.5 mg/L in ATCC 25923 and E19977 with pH 7.4, respectively.
  • Demonstrates therapeutic activity in a murine peritonitis-sepsis model, with all mice surviving at a dosage of 125 mg/kg via IV injection.